Fluorescein-12-dUTP 1 mM in TRIS Puffer (pH 7.5) (Perkin-Elmer)
Lieferant: AAT BIOQUEST
|
Achtung
|
17027
AATB17027EA
292
EUR
AATB17027
Fluorescein-12-dUTP 1 mM in TRIS Puffer (pH 7.5) (Perkin-Elmer)
Fluorescein-12-dUTP
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- Useful for DNA probe development
- Well-cited
- Multiple applications
Bitte beachten For research use only.
ADR:
5,II |