5-Brom-2'-deoxyuridin (Broxuridin) ≥95%

Lieferant: AAT BIOQUEST
Achtung

17030
AATB17030EA 120 EUR
AATB17030
5-Brom-2'-deoxyuridin (Broxuridin) ≥95%
5-Brom-2'-deoxyuridin (Broxuridin)

5-Bromo-2-deoxyuridine (BrdU) is an analog of thymidine, and can be incorporated into the newly synthesized DNA of replicating cells (S-phase cells).

Formel: C₉H₁₁BrN₂O₅
MDL: MFCD00006529
CAS-Nummer: 59-14-3
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR