5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP) 10 mM in Tris/EDTA (TE) Puffer ≥90%

Lieferant: AAT BIOQUEST
Achtung

17031
AATB17031EA 307 EUR
AATB17031
5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP) 10 mM in Tris/EDTA (TE) Puffer ≥90%
5-Bromo-2′-deoxyuridine-5′-triphosphate (5-Bromo-dUTP)

BrdUTP can be used for incorporating into DNA for the subsequent detection with anti-BrdU antibodies.

Formel: C₉H₁₁BrN₂Na₃O₁₄P₃
Molecular Weight: 612,99 g/mol
MDL: MFCD00057405
CAS-Nummer: 102212-99-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR