5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in Tris/EDTA (TE) Puffer ≥95%

Lieferant: AAT BIOQUEST
Achtung

17032
AATB17032EA 307 EUR
AATB17032
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in Tris/EDTA (TE) Puffer ≥95%
5-Bromouridine-5′-triphosphate (BrUTP )

BrUTP can be used for incorporating into RNA for the subsequent detection with anti-BrdU antibodies.

Formel: C₉H₁₁BrN₂Na₃O₁₅P₃
Molecular Weight: 628,98 g/mol
CAS-Nummer: 161848-60-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR