Cyanine 5-dATP [Cy5-dATP]

Lieferant: AAT BIOQUEST
Achtung

17038
AATB17038EA 143 EUR
AATB17038
Cyanine 5-dATP [Cy5-dATP]
Cyanine 5-dATP [Cy5-dATP]

The dye-modified deoxyadenosine 5'-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation, random prime labeling, cDNA labeling and 3'-end labeling.


  • Useful for DNA probe development
  • Well-cited
  • Multiple applications


Bitte beachten For research use only.

Molecular Weight: 1182,05 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR