iFluor® 488-dUTP 1 mM in TRIS Puffer (pH 7.5)

Lieferant: AAT BIOQUEST
Achtung

17039
AATB17039EA 137 EUR
AATB17039
iFluor® 488-dUTP 1 mM in TRIS Puffer (pH 7.5)
iFluor® 488-dUTP

The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • Useful for DNA probe development
  • Well-cited
  • Multiple applications


Bitte beachten For research use only.

Molecular Weight: 1152.83 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR