Aminopropargyl dCTP für PCR
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
5-Propargylamino-2'-deoxycytidine-5'-triphosphate
17050
AATB17050EA
769
EUR
AATB17050
Aminopropargyl dCTP für PCR
Aminopropargyl dCTP
The amine-modified deoxycytidine 5'-triphosphates (such as 5-propargylamino-2'-deoxycytidine-5'-triphosphate) can be used to produce amine-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- High and consistent DNA incorporation
- Efficient labeling
Molecular Weight:
543,95 g/mol |
CAS-Nummer:
2088422-92-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...