Aminopropargyl dUTP für PCR

Lieferant: AAT BIOQUEST
Achtung

17053
AATB17053EA 769 EUR
AATB17053
Aminopropargyl dUTP für PCR
Aminopropargyl dUTP

The amine-modified deoxyuridine 5'-triphosphates (such as 5-propargylamino-2'-deoxyuridine-5'-triphosphate) can be used to produce amine-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • High and consistent DNA incorporation
  • Efficient labeling

CAS-Nummer: 1380329-75-8
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a38567c1d2a15f1acf081c823fca54b40468a456&vid=383256490f02d21d69f54bc13ff2231f3c136ae3&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR