Aminopropargyl dATP für PCR
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
7-Deaza-7-Propargylamino-2'-deoxyadenosine-5'-triphosphate
17056
AATB17056EA
879
EUR
AATB17056
Aminopropargyl dATP für PCR
Aminopropargyl dATP
The amine-modified deoxyadenine 5'-triphosphates (such as 5-propargylamino-2'-deoxyadenine-5'-triphosphate) can be used to produce amine-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- High and consistent DNA incorporation
- Efficient labeling
Molecular Weight:
566,99 g/mol |
CAS-Nummer:
587848-72-4 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...