Aminopropargyl dGTP für PCR

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-Propargylamino-2'-deoxyguanosine-5'-triphosphate

17059
AATB17059EA 879 EUR
AATB17059
Aminopropargyl dGTP für PCR
Aminopropargyl dGTP

The amine-modified deoxyguanosine 5'-triphosphates (such as 5-propargylamino-2'-deoxyguanosine-5'-triphosphate) can be used to produce amine-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.


  • High and consistent DNA incorporation
  • Efficient labeling

Molecular Weight: 582,99 g/mol
CAS-Nummer: 587848-73-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87ca2f22e0f47783e0f2d0b2ae5e148da6b501fe&vid=c6c25881f69a1cd11c5bffd3bbd3643ad5cfca6e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR