XFD532 amine
Lieferant: AAT BIOQUEST
1706
AATB1706EA
309
EUR
AATB1706
XFD532 amine
XFD532 amine
XFD532 amine is the same molecule to the Alexa Fluor® 532 amine (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house