MagaDye™ 588 ddTTP
Lieferant: AAT BIOQUEST
17061 17065
AATB17061EA 1410 EUR
AATB17061 AATB17065
MagaDye™ 588 ddTTP
Nukleinsäure-Reagenzien Reagenzien für Sequentierung
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Bitte beachten For research use only.