Aminopropargyl ddCTP für die Sanger-Sequenzierung
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
[5-Propargylamino-2',3'-dideoxycytidine-5'-triphosphate
17070
AATB17070EA
1410
EUR
AATB17070
Aminopropargyl ddCTP für die Sanger-Sequenzierung
Aminopropargyl ddCTP
Sanger method is one of the most reliable and earliest DNA sequencing methods.
- Sanger sequencing building block
- Efficient labeling
Molecular Weight:
592,15 g/mol |
CAS-Nummer:
114748-56-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...