Aminopropargyl ddTTP für die Sanger-Sequenzierung

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-Propargylamino-2',3'-dideoxyuridine-5'-triphosphate

17072
AATB17072EA 1410 EUR
AATB17072
Aminopropargyl ddTTP für die Sanger-Sequenzierung
Aminopropargyl ddTTP

Sanger method is one of the most reliable and earliest DNA sequencing methods.


  • Sanger sequencing building block
  • Efficient labeling

Molecular Weight: 593,13 g/mol
CAS-Nummer: 114748-59-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=768836acebe11a000ae18ebed02732b3b9a0e802&vid=266e0388b0c08421400113c4e1c4e80811d53b6c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR