Aminopropargyl ddATP für die Sanger-Sequenzierung
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
7-Deaza-7-Propargylamino-2',3'-dideoxyadenosine-5'-triphosphate
17074
AATB17074EA
1410
EUR
AATB17074
Aminopropargyl ddATP für die Sanger-Sequenzierung
Aminopropargyl ddATP
Sanger method is one of the most reliable and earliest DNA sequencing methods.
- Sanger sequencing building block
- Efficient labeling
Molecular Weight:
615,19 g/mol |
CAS-Nummer:
114748-69-5 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...