Aminopropargyl ddGTP für die Sanger-Sequenzierung
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
7-Deaza-7-Propargylamino-2',3'-dideoxyguanosine-5'-triphosphate
17076
AATB17076EA
2150
EUR
AATB17076
Aminopropargyl ddGTP für die Sanger-Sequenzierung
Aminopropargyl ddGTP
Sanger method is one of the most reliable and earliest DNA sequencing methods.
- Sanger sequencing building block
- Efficient labeling
Molecular Weight:
631,19 g/mol |
CAS-Nummer:
114748-61-7 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...