XFD568 amine
Lieferant: AAT BIOQUEST
1708
AATB1708EA
309
EUR
AATB1708
XFD568 amine
XFD568 amine
XFD568 amine is the same molecule to the Alexa Fluor® 568 amine (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only