2-Aminoethoxypropargyl ddATP (1 µmole)
Lieferant: AAT BIOQUEST
17084
AATB17084EA
1410
EUR
AATB17084
2-Aminoethoxypropargyl ddATP (1 µmole)
Nukleinsäure-Reagenzien
Reagenzien für Sequentierung
Sanger method is one of the most reliable and earliest DNA sequencing methods.
- In-house quality control
- Useful for sanger sequencing applications
Bitte beachten For research use only.