XFD647 amine
Lieferant: AAT BIOQUEST
1709
AATB1709EA
309
EUR
AATB1709
XFD647 amine
XFD647 amine
XFD647 amine is the same molecule to the Alexa Fluor® 647 amine (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only