7-Deaza-7-Propargylamino-3'-azidomethyl-dATP

Lieferant: AAT BIOQUEST
Achtung

17090
AATB17090EA 586 EUR
AATB17090
7-Deaza-7-Propargylamino-3'-azidomethyl-dATP
7-Deaza-7-Propargylamino-3'-azidomethyl-dATP

7-Deaza-7-Propargylamino-3'-azidomethyl-dATP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).


  • Ngs building block
  • Useful for multiple applications
  • High quality


Bitte beachten For research use only.

Formel: C15H21N8O12P3
Molecular Weight: 598,3 g/mol
CAS-Nummer:  666847-93-4
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR