5-Propargylamino-3'-azidomethyl-dCTP

Lieferant: AAT BIOQUEST
Achtung

17091
AATB17091EA 367 EUR
AATB17091
5-Propargylamino-3'-azidomethyl-dCTP
5-Propargylamino-3'-azidomethyl-dCTP

5-Aminopropargyl 3’-azidomethyl dCTP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).


  • Ngs building block
  • Useful for multiple applications
  • High quality


Bitte beachten For research use only.

Formel: C13H20N7O13P3
Molecular Weight: 575,26 g/mol
CAS-Nummer:  666847-71-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR