7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP

Lieferant: AAT BIOQUEST
Achtung

17092
AATB17092EA 513 EUR
AATB17092
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP
7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP

7-Deaza-7-Propargylamino-3'-azidomethyl-dGTP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).


  • Ngs building block
  • Useful for multiple applications
  • High quality


Bitte beachten For research use only.

Formel: C15H21N8O13P3
Molecular Weight: 614,3 g/mol
CAS-Nummer: 666847-81-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR