5-Propargylamino-3'-azidomethyl-dUTP
Lieferant: AAT BIOQUEST
|
Achtung
|
17093
AATB17093EA
367
EUR
AATB17093
5-Propargylamino-3'-azidomethyl-dUTP
5-Propargylamino-3'-azidomethyl-dUTP
5-Propargylamino-3'-azidomethyl-dUTP is a key building block for preparing fluorescent conjugates that are used in the next generation sequencing (NGS).
- Ngs building block
- Useful for multiple applications
- High quality
Bitte beachten For research use only.
Formel:
C13H19N6O14P3 Molecular Weight: 576,24 g/mol |
CAS-Nummer:
666847-57-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...