cAMP-NHS ester

Lieferant: AAT BIOQUEST
Achtung

17102
AATB17102EA 769 EUR
AATB17102
cAMP-NHS ester
cAMP-NHS ester

Cyclic adenosine monophosphate (cAMP) is a second messenger used for intracellular signal induction.


  • Camp probe/assay building block
  • Useful for investigating cellular behavior


Bitte beachten For research use only.

Molecular Weight: 684.64 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR