ReadiUse™ dNTP Mix *10 mM* (5 ml)
Lieferant: AAT BIOQUEST
17200
AATB17200EA 439 EUR
AATB17200
ReadiUse™ dNTP Mix *10 mM* (5 ml)
Nukleinsäure-Reagenzien Nukleotide
Our Ready-to Use Solution of deoxynucleotide triphosphates (dNTPs) have been extensively tested and verified for use in a wide variety of molecular biology applications, including PCR, real-time PCR, high-fidelity PCR, long PCR, cDNA synthesis, RT-PCR, RCA, MDA, DNA labelling, DNA sequencing, in vitro transcription, siRNA synthesis, and RNA amplification.
  • Ready to use formulation
  • In-house testing and quality control
  • Multiple applications
Bitte beachten For research use only.