ReadiUse™ dNTP Mix *10 mM* (5 ml)
Lieferant: AAT BIOQUEST
17200
AATB17200EA
439
EUR
AATB17200
ReadiUse™ dNTP Mix *10 mM* (5 ml)
Nukleinsäure-Reagenzien
Nukleotide
Our Ready-to Use Solution of deoxynucleotide triphosphates (dNTPs) have been extensively tested and verified for use in a wide variety of molecular biology applications, including PCR, real-time PCR, high-fidelity PCR, long PCR, cDNA synthesis, RT-PCR, RCA, MDA, DNA labelling, DNA sequencing, in vitro transcription, siRNA synthesis, and RNA amplification.
- Ready to use formulation
- In-house testing and quality control
- Multiple applications
Bitte beachten For research use only.