ReadiUse™ PCR Reaction Buffer
Lieferant: AAT BIOQUEST
17202
AATB17202EA
133
EUR
AATB17202
ReadiUse™ PCR Reaction Buffer
Puffer
Assay-Puffer
This ready to use PCR buffer contains several components essential for the PCR reaction.
- Designed for ease of use
- MgCl2 (magnesium chloride), a cofactor, is used for maintaining the activity of the DNA polymerase enzyme
PCR (polymerase chain reaction) is a laboratory technique used to amplify DNA (deoxyribonucleic acid) segments.
Bitte beachten For research use only.