ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Lieferant: AAT BIOQUEST
17310
AATB17310EA 682 EUR
AATB17310
ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Proteinkonjugate und Markierungen
The kit provides Taq DNA polymerase enzyme, which incorporates iFluor® 488-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
  • Robust protocol
  • Bright labelling
  • Photostable fluorescence
The Readilink™ iFluor® 488 FISH fluorescence imaging kit is a convenient tool for labelling a target DNA using an iFluor® 488 labeled FISH probe via in situ hybridisation.
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
Verpackung: Packaged in plastic vials.
Bitte beachten For research use only.