ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Lieferant: AAT BIOQUEST
The kit provides Taq DNA polymerase enzyme, which incorporates iFluor® 488-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
- Robust protocol
- Bright labelling
- Photostable fluorescence
The Readilink™ iFluor® 488 FISH fluorescence imaging kit is a convenient tool for labelling a target DNA using an iFluor® 488 labeled FISH probe via in situ hybridisation.
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
Verpackung: Packaged in plastic vials.
Bitte beachten For research use only.