ReadiLink™ Cy3 Nick Translation dsDNA Labelling Kit
Lieferant: AAT BIOQUEST
17406 17407
AATB17406EA 544 EUR
AATB17406 AATB17407
ReadiLink™ Cy3 Nick Translation dsDNA Labelling Kit
Proteinkonjugate und Markierungen
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearised plasmid DNA.
  • Provides all necessary reagents
  • Adjustable ratios
  • Wide sample type compatibility
The resulted Cy3-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridisation (FISH)..
Verpackung: Packaged in plastic vials.
Bitte beachten For research use only.