Acridinorange 10 mg/ml ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
Brilliant Acridine Orange E, 3,6-Bis(dimethylamino)acridine, Acridine Orange base, Solvent Orange 15, 3,6-Bis(dimethylamino)acridine N,N,N',N'-Tetramethyl-3,6-acridinediamine, Acridine orange free base, Acridine Orange Stain
17503
AATB17503EA
120
EUR
AATB17503
Acridinorange 10 mg/ml ≥95%
Acridinorange
Acridine orange is a nucleic acid selective fluorescent cationic dye useful for cell cycle determination.
- Binding-Determined Bicolor Fluorescence
- Cell-Permeable
Bitte beachten For research use only.
Formel:
C₁₇H₁₉N₃ Molecular Weight: 265,35 g/mol |
MDL:
MFCD00005029 CAS-Nummer: 494-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...