Actinomycin D antibiotic

Lieferant: AAT BIOQUEST
Achtung

17505
AATB17505EA 120 EUR
AATB17505
Actinomycin D antibiotic
Actinomycin D

Actinomycin is a polypeptide antibiotic isolated from soil bacteria of the genus Streptomyces.


  • Well-researched
  • Traditional chemotherapy drug

Formel: C₆₂H₈₆N₁₂O₁₆
MDL: MFCD00005033
CAS-Nummer: 50-76-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=babc4e00a60c47846c881ad19b4b58158ffacf38&vid=ba9a333f02a8b43d5f2f8d924c33b1d252eeb157&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR