4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 10 mM in Wasser ≥95%

Lieferant: AAT Bioquest

17507
AATB17507EA 120 EUR
AATB17507
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 10 mM in Wasser ≥95%
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

DAPI is a fluorescent stain that binds strongly to DNA.


  • Binding-Determined Bicolor Fluorescence
  • Cell-Permeable
  • Useful For Multicolor Imaging


Bitte beachten For research use only.

Formel: C₁₆H₁₅N₅·2HCl
MDL: MFCD00012681
CAS-Nummer: 28718-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR