4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) ≥95%

Lieferant: AAT Bioquest
Achtung

17510 17511 17513
AATB17510EA 120 EUR
AATB17510 AATB17511 AATB17513
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) ≥95%
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

DAPI is a fluorescent stain that binds strongly to DNA.

Formel: C₁₆H₁₅N₅·2HCl
MDL: MFCD00012681
CAS-Nummer: 28718-90-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88ffc6bce3cf6f3eb043b711207f00f128de1153&vid=ee288a4b4ed9f0d89ef95afc438b50ef245efa2f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR