4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) ≥95%
Lieferant: AAT Bioquest
|
Achtung
|
17510
17511
17513
AATB17510EA
120
EUR
AATB17510
AATB17511
AATB17513
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) ≥95%
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)
DAPI is a fluorescent stain that binds strongly to DNA.
Formel:
C₁₆H₁₅N₅·2HCl |
MDL:
MFCD00012681 CAS-Nummer: 28718-90-3 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...