Hydroxystilbamidine bis(methanesulphonate) ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Fluoro-Gold™

17514
AATB17514EA 152 EUR
AATB17514
Hydroxystilbamidine bis(methanesulphonate) ≥95%
Hydroxystilbamidine bis(methanesulphonate)

Hydroxystilbamidine (also called Fluoro Gold) is used for staining DNA and RNA.

Formel: C₁₈H₂₄N₄O₇S₂
Molecular Weight: 472,54 g/mol
MDL: MFCD01863083
CAS-Nummer: 223769-64-0
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR