Thiazole orange ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 1-Methyl-4-[(3-methyl-2(3H)-benzothiazolylidene)methyl]quinolinium p-tosylate

17518
AATB17518EA 307 EUR
AATB17518
Thiazole orange ≥90%
Thiazole orange

Thiazole orange is a fluorescent DNA-binding dye for reticulocyte analysis.

Formel: C₂₆H₂₄N₂O₃S₂
Molecular Weight: 476,61 g/mol
MDL: MFCD00192499
CAS-Nummer: 107091-89-4
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR