bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 20 mM in Wasser ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
Bisbenzimide H 33258, 4-(5-(4-Methylpiperazin-1-yl)-1H,1'H-[2,5'-bibenzo[d]imidazol]-2'-yl)phenol trihydrochloride, 2'-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bibenzimidazole trihydrochloride hydrate
17525
AATB17525EA
152
EUR
AATB17525
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) 20 mM in Wasser ≥95%
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.
- Bright Fluorescence
- 95% Purity
- Useful For Labeling Nuclei/Mitochondria
- DAPI Alternative
Bitte beachten For research use only.
Formel:
C₂₅H₂₄N₆O·3HCl·xH₂O Molecular Weight: 533,88 g/mol (anhy) |
MDL:
MFCD00012679 CAS-Nummer: 23491-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...