bisBenzimide H-33342 (Hoechst 33342) 20 mM in Wasser ≥95%

Lieferant: AAT BIOQUEST

Synonyme: Hoechst 33342

17535
AATB17535EA 152 EUR
AATB17535
bisBenzimide H-33342 (Hoechst 33342) 20 mM in Wasser ≥95%
bisBenzimide H-33342 (Hoechst 33342)

The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.


  • Bright Fluorescence
  • 95% Purity
  • Useful For Labeling Nuclei/Mitochondria
  • DAPI Alternative


Bitte beachten For research use only.

Formel: C₂₇H₂₈N₆O
Molecular Weight: 452,562 g/mol
CAS-Nummer: 23491-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR