Hoechst 34580 trihydrochloride 20 mM in Wasser ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
N,N-Dimethyl-4-(6-(4-methylpiperazin-1-yl)-1H,3'H-[2,5'-bibenzo[d]imidazol]-2'-yl)aniline trihydrochloride, Hoechst 34580.3HCl, HOE 34580.3HCl
17538
AATB17538EA
307
EUR
AATB17538
Hoechst 34580 trihydrochloride 20 mM in Wasser ≥95%
Hoechst 34580 trihydrochloride
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.
- Bright Fluorescence
- 95% Purity
- Useful For Labeling Nuclei/Mitochondria
- DAPI Alternative
Bitte beachten For research use only.
Formel:
C₂₇H₂₉N₇ . 3HCl Molecular Weight: 560,95 g/mol |
CAS-Nummer:
911004-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...