DRAQ5 5 mM water, ReadiUse™ staining solution

Lieferant: AAT BIOQUEST

17558
AATB17558EA 232 EUR
AATB17558
DRAQ5 5 mM water, ReadiUse™ staining solution
DRAQ5

DRAQ5™ is the trade name of 1,5-bis{[2-(di-methylamino) ethyl]amino}-4, 8-dihydroxyanthracene-9,10-dione (DRAQ-5™ is the trade mark of Biostatus Ltd).


  • Stable for 12 months


Quality tested in-house


Bitte beachten For Research Use Only

Molecular Weight: 485.41 g/mol
CAS-Nummer: 252903-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR