Nuclear Green™ Fixable DCS1 5 mM in Wasser
Lieferant: AAT BIOQUEST
17569
AATB17569EA 402 EUR
AATB17569
Nuclear Green™ Fixable DCS1 5 mM in Wasser
Nuclear Green™ Fixable DCS1
Nuclear Green™ Fixable DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only