Nuclear Green™ Fixable DCS1 5 mM in Wasser
Lieferant: AAT BIOQUEST
Nuclear Green™ Fixable DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only