Thiazole orange homodimer (TOTO®-1) 5 mM in DMSO ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
DiTO™-1, TOTO-1, TOhD
17575
AATB17575EA
307
EUR
AATB17575
Thiazole orange homodimer (TOTO®-1) 5 mM in DMSO ≥95%
Thiazole orange homodimer (TOTO®-1)
DiTO™-1 is chemically equivalent to TOTO®-1 (TOTO® is the trademark of Invitrogen).
Formel:
C₄₉H₅₈I₄N₆S₂ Molecular Weight: 1303 g/mol |
CAS-Nummer:
143413-84-7 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...