DiYO™-3 5 mM in DMSO ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
YOYO®-3
17581
AATB17581EA
307
EUR
AATB17581
DiYO™-3 5 mM in DMSO ≥90%
DiYO™-3
DiYO™-3 is chemically equivalent to YOYO®-3 (YOYO® is the trademark of Invitrogen).
Formel:
C₅₃H₅₈I₄N₆O₂ Molecular Weight: 1322.73 g/mol |
CAS-Nummer:
156312-20-8 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...