Propidiumiodid 1 mg/ml in wässriger Lösung

Lieferant: AAT BIOQUEST

Synonyme: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid

17586 17585
AATB17586EA 135 EUR
AATB17586 AATB17585
Propidiumiodid 1 mg/ml in wässriger Lösung
Propidiumiodid

Propidium iodide (PI) is a red-fluorescent dye widely used for staining and identifying dead cell populations in flow cytometry and fluorescence microscopy.


  • Stable for 6 months


PCR (polymerase chain reaction) is a laboratory technique used to amplify DNA (deoxyribonucleic acid) segments.


Bitte beachten For Research Use Only

Formel: C₂₇H₃₄I₂N₄
MDL: MFCD00011921
CAS-Nummer: 25535-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR