Gelite™ Green Nucleic Acid Gel Staining Kit
Lieferant: AAT BIOQUEST
17589
AATB17589EA
307
EUR
AATB17589
Gelite™ Green Nucleic Acid Gel Staining Kit
Tests
Nukleinsäure-Tests
Gelite™ Green is a sensitive fluorescent nucleic acid gel stain for detecting nucleic acids in agarose and polyacrylamide gels.
- 2× more sensitive than etbr
- Brighter signal
- Exceptional DNA affinity
- Robust protocol
- Multiple applications
Bitte beachten For research use only.