Helixyte™ Green *20X Aqueous PCR Solution*
Lieferant: AAT BIOQUEST
17591
AATB17591EA 307 EUR
AATB17591
Helixyte™ Green *20X Aqueous PCR Solution*
Tests qPCR-Assays
Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).
  • Same spectral properties to SYBR® gold
  • High sensitivity
  • Useful for nucleic acid labeling in gels
Bitte beachten For research use only.