Gelite™ Orange Nucleic Acid Gel Staining Kit
Lieferant: AAT BIOQUEST
Gelite™ Orange is an extremely sensitive nucleic acid gel stain for detecting DNA or RNA in gels using a standard 300 nm UV transilluminator and Polaroid 667 black-and-white print film.
- High sensitivity
- Exceptional DNA affinity
- Robust protocol
- Multiple applications
Bitte beachten For research use only.