Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml)
Lieferant: AAT BIOQUEST
17595
AATB17595EA
460
EUR
AATB17595
Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml)
Färbemittel
Elektrophorese-Färbemittel
Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).
- Same spectral properties to SYBR® gold
- High sensitivity
- Useful for nucleic acid labeling in gels
Bitte beachten For research use only.