Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml)
Lieferant: AAT BIOQUEST
17595
AATB17595EA 460 EUR
AATB17595
Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml)
Färbemittel Elektrophorese-Färbemittel
Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).
  • Same spectral properties to SYBR® gold
  • High sensitivity
  • Useful for nucleic acid labeling in gels
Bitte beachten For research use only.