Helixyte™ Green Nucleic Acid Gel Stain 10,000X DMSO Solution
Lieferant: AAT Bioquest
17590
17604
AATB17590EA
769
EUR
AATB17590
AATB17604
Helixyte™ Green Nucleic Acid Gel Stain 10,000X DMSO Solution
Färbemittel
Elektrophorese-Färbemittel
Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).
- Same spectral properties to SYBR® green
- High sensitivity
- Useful for nucleic acid labeling in gels
Bitte beachten For research use only.