Helixyte™ Green Nucleic Acid Gel Stain 10,000X DMSO Solution
Lieferant: AAT Bioquest
17590 17604
AATB17590EA 769 EUR
AATB17590 AATB17604
Helixyte™ Green Nucleic Acid Gel Stain 10,000X DMSO Solution
Färbemittel Elektrophorese-Färbemittel
Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).
  • Same spectral properties to SYBR® green
  • High sensitivity
  • Useful for nucleic acid labeling in gels
Bitte beachten For research use only.