Helixyte™ Green ssDNA-Reagenz
Lieferant: AAT BIOQUEST
Helixyte™ Green ssDNA Quantifying Reagent is an excellent alternative for quantifying ssDNA and oligonucleotides with significantly improved sensitivity and selectivity.
- Stable for 12 months
Synthetic oligonucleotides are used in several molecular biology techniques, such as DNA sequencing, site-directed mutagenesis, DNA amplification, and in situ hybridization.
Bitte beachten For Research Use Only