Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Lieferant: AAT BIOQUEST
17623
AATB17623EA 281 EUR
AATB17623
Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Tests Nukleinsäure-Tests
Helixyte™ Green fluorimetric ssDNA quantitation kit is optimised for quantifying dsDNA with a fluorescence microplate reader.
  • Optimized for microplate readers
  • High sensitivity
  • Low interference from common contaminants
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution. It enables researchers to quantify as little as 100 pg/ml oligonucleotide or ssDNA. This sensitivity exceeds that achieved with absorbance methods by more than 10000-fold.
With this kit, as little as 1 ng/ml oligonucleotide or ssDNA might be detected.
Verpackung: Plastic vial.
Bitte beachten For research use only.